Phenyl-4-hydroxybenzoate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P1680.10 P1680.50
XXXXP1680.10EA 0 GBP
XXXXP1680.10 XXXXP1680.50
Phenyl-4-hydroxybenzoate
Phenyl 4-hydroxybenzoate
Formula: C₁₃H₁₀O₃
MDL Number: MFCD00016467
CAS Number: 17696-62-7
EINECS: 241-698-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR