Di-n-octyl phthalate ≥98.0% (by GC), Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIAL80153-100MLEA
225
GBP
SIAL80153-100ML
Di-n-octyl phthalate ≥98.0% (by GC), Sigma-Aldrich®
Di-n-octyl phthalate
Formula:
C₂₄H₃₈O₄ MW: 390.56 g/mol Density: 0.980 g/mL (20 °C) (lit.) Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015292 CAS Number: 117-84-0 EINECS: 204-214-7 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...