5-Methyl-2,1,3-benzothiadiazole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

017298-5G 017298-10G 017298-25G
FLUO017298-5GEA 0 GBP
FLUO017298-5G FLUO017298-10G FLUO017298-25G
5-Methyl-2,1,3-benzothiadiazole
5-Methyl-2,1,3-benzothiadiazole
Formula: C₇H₆N₂S
MDL Number: MFCD00173945
CAS Number: 1457-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR