4-Amino-2-chloro-6,7-dimethoxyquinazoline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Doxazosin Related Compound C, 2-Chloro-4-amino-6,7-dimethoxyquinazoline

024183-25G 024183-5G
FLUO024183-25GEA 0 GBP
FLUO024183-25G FLUO024183-5G
4-Amino-2-chloro-6,7-dimethoxyquinazoline
4-Amino-2-chloro-6,7-dimethoxyquinazoline
Formula: C₁₀H₁₀ClN₃O₂
MW: 239.66 g/mol
MDL Number: MFCD00051734
CAS Number: 23680-84-4
EINECS: 245-821-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR