4,7-Dibromo-2,1,3-benzothiadiazole, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
693847-1G
693847-5G
SIAL693847-1GEA
54.3
GBP
SIAL693847-1G
SIAL693847-5G
4,7-Dibromo-2,1,3-benzothiadiazole, Sigma-Aldrich®
4,7-Dibromo-2,1,3-benzothiadiazole
Formula:
C₆H₂Br₂N₂S MW: 293.97 g/mol |
MDL Number:
MFCD00658844 CAS Number: 15155-41-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...