6-Morpholinonicotinic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7747-1G
PY-7747-5G
COBBPY-7747-1GEA
0
GBP
COBBPY-7747-1G
COBBPY-7747-5G
6-Morpholinonicotinic acid
6-Morpholinonicotinic acid
Formula:
C₁₀H₁₂N₂O₃ MW: 208.213 g/mol |
MDL Number:
MFCD02682034 CAS Number: 120800-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...