Potassium-4-(trifluoromethyl)phenyltrifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Potassium 4-(trifluoromethyl)phenyltrifluoroborate, (T-4)-Potassium trifluoro(4-(trifluoromethyl)phenyl)borate

TB-3217-1G TB-3217-25G TB-3217-5G
COBBTB-3217-1GEA 0 GBP
COBBTB-3217-1G COBBTB-3217-25G COBBTB-3217-5G
Potassium-4-(trifluoromethyl)phenyltrifluoroborate
Potassium 4-(trifluoromethyl)phenyltrifluoroborate
Formula: C₇H₄BF₆K
MW: 252.01 g/mol
MDL Number: MFCD04039955
CAS Number: 166328-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR