Potassium-4-methoxy-2-methylphenyltrifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TB-3239-25G TB-3239-1G TB-3239-5G
COBBTB-3239-25GEA 0 GBP
COBBTB-3239-25G COBBTB-3239-1G COBBTB-3239-5G
Potassium-4-methoxy-2-methylphenyltrifluoroborate
Potassium 4-methoxy-2-methylphenyltrifluoroborate
Formula: C₈H₉BF₃KO
MDL Number: MFCD04115764
CAS Number: 850623-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR