2-(Hydroxymethyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-(Hydroxymethyl)phenyl)boronic acid, 2-Hydroxymethylphenylboronic acid, 2-(Hydroxymethyl)benzeneboronic acid

BB-8356-5G BB-8356-1G BB-8356-25G
COBBBB-8356-5GEA 31.66 GBP
COBBBB-8356-5G COBBBB-8356-1G COBBBB-8356-25G
2-(Hydroxymethyl)phenylboronic acid
2-(Hydroxymethyl)phenylboronic acid
Formula: C₇H₉BO₃
MW: 151.96 g/mol
MDL Number: MFCD02258966
CAS Number: 87199-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR