Bis(2-methoxyethyl)phthalate
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Bis(2-methoxyethyl) phthalate
EHERC16174400EA
94.4
GBP
EHERC16174400
Bis(2-methoxyethyl)phthalate
Bis(2-methoxyethyl) phthalate
Formula:
C₁₄H₁₈O₆ MW: 282.29 g/mol |
MDL Number:
MFCD00042842 CAS Number: 117-82-8 EINECS: 204-212-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...