Methyl thiosalicylate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-mercaptobenzoate

008066-5G 008066-25G
FLUO008066-5GEA 0 GBP
FLUO008066-5G FLUO008066-25G
Methyl thiosalicylate
Methyl thiosalicylate
Formula: C₈H₈O₂S
MW: 168.21 g/mol
MDL Number: MFCD00060678
CAS Number: 4892-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR