3-Bromo-5-fluorobenzhydrazide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4724-5G CA-4724-1G CA-4724-250M
COBBCA-4724-5GEA 0 GBP
COBBCA-4724-5G COBBCA-4724-1G COBBCA-4724-250M
3-Bromo-5-fluorobenzhydrazide
3-Bromo-5-fluorobenzhydrazide
Formula: C₇H₆BrFN₂O
MDL Number: MFCD11619361
CAS Number: 1094510-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR