3-(3-Fluoro-4-methoxybenzoyl)propionic acid

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

F1368.5 F1368.25
XXXXF1368.5EA 0 GBP
XXXXF1368.5 XXXXF1368.25
3-(3-Fluoro-4-methoxybenzoyl)propionic acid
3-(3-Fluoro-4-methoxybenzoyl)propionic acid
Formula: C₁₁H₁₁FO₄
MW: 226.20 g/mol
MDL Number: MFCD00020538
CAS Number: 347-63-7
EINECS: 206-474-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR