(2-Chloro-1,3-thiazol-5-yl)methanol

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Chlorothiazol-5-yl)methanol

7N-099.500 7N-099.1 7N-099.5 7N-099.10
BIOT7N-099.500EA 0 GBP
BIOT7N-099.500 BIOT7N-099.1 BIOT7N-099.5 BIOT7N-099.10
(2-Chloro-1,3-thiazol-5-yl)methanol
(2-Chloro-1,3-thiazol-5-yl)methanol
Formula: C₄H₄NOClS
MW: 149.6000 g/mol
MDL Number: MFCD03001277
CAS Number: 145015-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR