2-Hydrazinobenzothiazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Hydrazinylbenzo[d]thiazole

M-Clarity™ quality level = MQ100

642436-25G 642436-5G
SIAL642436-25GEA 345 GBP
SIAL642436-25G SIAL642436-5G
2-Hydrazinobenzothiazole, Sigma-Aldrich®
2-Hydrazinobenzothiazole
Formula: C₇H₇N₃S
MW: 165.21 g/mol
MDL Number: MFCD00041849
CAS Number: 615-21-4
EINECS: 210-416-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR