3-Bromo-1-(trimethylsilyl)-1-propyne

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Trimethylsilyl)propargyl bromide, (3-Bromoprop-1-yn-1-yl)trimethylsilane

SILS03140.1 SILS03140.5
SILNSILS03140.1EA 0 GBP
SILNSILS03140.1 SILNSILS03140.5
3-Bromo-1-(trimethylsilyl)-1-propyne
3-Bromo-1-(trimethylsilyl)-1-propyne
Formula: C₆H₁₁BrSi
MW: 191.14 g/mol
MDL Number: MFCD00134460
CAS Number: 38002-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR