5-Bromo-4-(4-methoxyphenyl)pyrimidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6124-5G HC-6124-1G
COBBHC-6124-5GEA 0 GBP
COBBHC-6124-5G COBBHC-6124-1G
5-Bromo-4-(4-methoxyphenyl)pyrimidine
5-Bromo-4-(4-methoxyphenyl)pyrimidine
Formula: C₁₁H₉BrN₂O
MDL Number: MFCD09800946
CAS Number: 941294-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR