Chlorobis(3,5-dimethyl-4-methoxyphenyl)phosphine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

695130-100MG 695130-500MG
SIAL695130-100MGEA 115 GBP
SIAL695130-100MG SIAL695130-500MG
Chlorobis(3,5-dimethyl-4-methoxyphenyl)phosphine, Sigma-Aldrich®
Chlorobis(3,5-dimethyl-4-methoxyphenyl)phosphine
Formula: C₁₈H₂₂ClO₂P
MDL Number: MFCD01630810
CAS Number: 136802-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR