Cyclohexyldiphenylphosphine ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Diphenylcyclohexylphosphine
36708.03EA
36.3
GBP
36708.03
36708.14
36708.22
36708.06
Cyclohexyldiphenylphosphine ≥98%
Cyclohexyldiphenylphosphine
Formula:
C₁₈H₂₁P MW: 268.33 g/mol Melting Pt: 58…62 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00046360 CAS Number: 6372-42-5 EINECS: 228-904-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...