Estriol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

SIALE1149-1GEA 445 GBP
SIALE1149-1G
Estriol, Sigma-Aldrich®
Estriol
Formula: C₁₈H₂₄O₃
MDL Number: MFCD00003691
CAS Number: 50-27-1
EINECS: 200-022-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR