Estriol, Supelco®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
46565-100MGEA
72.4
GBP
46565-100MG
Estriol, Supelco®
Estriol
Formula:
C₁₈H₂₄O₃ |
MDL Number:
MFCD00003691 CAS Number: 50-27-1 EINECS: 200-022-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...