Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')scandium(III) ≥99% (99.9%-Sc)

Supplier: Strem Chemicals, Inc.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Sc(TMHD)3Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)scandium(III)

STRE21-1000-500MGEA 0 GBP
STRE21-1000-500MG
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')scandium(III) ≥99% (99.9%-Sc)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')scandium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇O₆Sc
MW: 594.758 g/mol
Boiling Pt: 275 °C (dec.)
Melting Pt: 150…152 °C
MDL Number: MFCD00169875
CAS Number: 15492-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR