Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Al(TMHD)3
, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)aluminum
M-Clarity™ quality level = MQ100
SIAL397288-5GEA
435
GBP
SIAL397288-5G
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium, Sigma-Aldrich®
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')aluminium
Formula:
C₃₃H₅₇AlO₆ MW: 576.78 g/mol |
MDL Number:
MFCD00058918 CAS Number: 14319-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...