Anti-HLA-DR+DP Mouse Monoclonal Antibody [clone: 6D461]

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H6098-72D
USBIH6098-72DEA 507 GBP
USBIH6098-72D
Anti-HLA-DR+DP Mouse Monoclonal Antibody [clone: 6D461]
Antibodies
Type: Primary
Antigen: HLA-DR+DP
Clonality: monoclonal
Clone: 6D461
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol HLA-DR DP
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 6D461
Reactivity Human
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoPrecipitation Yes
Flow cytometry Yes
Form liquid
Purification purified by Protein A chromatography
Concentration ~0,5 mg/ml
Storage buffer PBS, pH 7,4, 15mM sodium azide, before the addition of glycerol to 40%.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR