Ethidium monoazide bromide (EMA) ≥95% (by HPLC), Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

E2028-5MG
E2028-5MGEA 424 GBP
E2028-5MG
Ethidium monoazide bromide (EMA) ≥95% (by HPLC), Sigma-Aldrich®
Ethidium monoazide bromide (EMA)

Ethidium bromide monoazide. ≥95% (HPLC), solid (3-Amino-8-azido-5-ethyl-6-phenylphenanthridinium bromide; Ethidium monoazide bromide)

Formula: C₂₁H₁₈BrN₅
MW: 420.305 g/mol
Storage Temperature: Freezer
CAS Number: 58880-05-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=abb8b4cb6758532ba8a5437570882449ace59c39&vid=83312ded18e693eaa02db2807f0137a027b82c4f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR