Anti-GABRP/GABA A receptor pi Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-13257R-A680
BOSSBS-13257R-A680EA 835 GBP
BOSSBS-13257R-A680
Anti-GABRP/GABA A receptor pi Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
GAD-65 and GAD-67, glutamate decarboxylases function to catalyze the production of GABA (Gamma-aminobutyric acid). In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl-conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. Both GABAA and GABAC are ligand-gated ion channels, however, they are structurally and functionally distinct.

Type: Primary
Antigen: GABRP/GABA A receptor pi
Clonality: Polyclonal
Clone:
Conjugation:More Product Information
Order Now

SPECIFICATIONS

Antigen name GABRP/GABA A receptor pi
Antigen synonyms GABAA receptor|GABRP|Gamma aminobutyric acid receptor pi subunit|MGC126386|Gamma aminobutyric acid GABA A receptor pi|MGC126387|Gamma-aminobutyric acid receptor subunit pi|Gamma aminobutyric acid receptor subunit pi|GBRP_HUMAN|Gamma aminobutyric acid A receptor pi|GABAA receptor subunit pi
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 680
Reactivity Mouse
Host Rabbit
Gene ID 2568
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Unmodified
Epitope 81-180/440
Immunogen KLH conjugated synthetic peptide derived from human GABRP/GABA A receptor pi
Form Liquid
Molecular weight 49 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
Storage temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR