Rizatriptan benzoate

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: N,N-Dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethylamine Benzoate

462940010
ACRO462940010EA 179 GBP
ACRO462940010
Rizatriptan benzoate
Rizatriptan benzoate

  • Bioactive

Formula: C₂₂H₂₅N₅O₂
MW: 391.475 g/mol
Melting Pt: 178…180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866224
CAS Number: 145202-66-0

Order Now

Specification Test Results

Appearance White to beige Powder or crystals
Infrared spectrum Conforms
Melting point 178 to 182 °C
Water ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR