GSK1120212 98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N-(3-(3-Cyclopropyl-5-((2-fluoro-4-iodophenyl)amino)-6,8-dimethyl-2,4,7-trioxo-3,4,6,7-tetrahydropyrido[4,3-d]pyrimidin-1(2H)-yl)phenyl)acetamide, Trametinib, JTP-74057

473411000
473411000.EA 148 GBP
473411000.
GSK1120212 98%
GSK1120212
Formula: C₂₆H₂₃FIN₅O₄
MW: 615.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17215075
CAS Number: 871700-17-3

Order Now

Specification Test Results

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR