Methanol analytical reagent

Supplier: PROLABO CMP
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Hydroxy methaneMethyl alcoholCarbinolWood alcohol

PRLS88893.290EA 0 GBP
PRLS88893.290
Methanol analytical reagent
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://uk.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR