3,4-Difluorophenol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002091-25G
002091-100G
002091-10G
FLUO002091-25GEA
0
GBP
FLUO002091-25G
FLUO002091-100G
FLUO002091-10G
3,4-Difluorophenol
3,4-Difluorophenol
Formula:
C₆H₄F₂O Boiling Pt: 185 °C (1013 hPa) Melting Pt: 32…36 °C |
MDL Number:
MFCD00010315 CAS Number: 2713-33-9 UN: 2921 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...