p-Anisic acid

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Methoxybenzoic acid4-Anisic acid

EHERC10265900EA 77.9 GBP
EHERC10265900
p-Anisic acid
p-Anisic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 182…185 °C
Density: ∼1.385 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002542
CAS Number: 100-09-4
EINECS: 202-818-5
Merck Index: 13,00669

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR