Thiocarbonyl chloride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Thiophosgene
M-Clarity™ quality level = MQ200
115150-100G
115150-25G
115150-5G
SIAL115150-100GEA
198
GBP
SIAL115150-100G
SIAL115150-25G
SIAL115150-5G
Thiocarbonyl chloride, Sigma-Aldrich®
Thiocarbonyl chloride
Formula:
CCl₂S |
MDL Number:
MFCD00004918 CAS Number: 463-71-8 EINECS: 207-341-6 UN: 2474 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...