2,2,3,3,4,4,5,5-Octafluoro-1-pentanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1H,1H,5H-Octafluoro-1-pentanol

M-Clarity™ quality level = MQ100

SIAL269433-100GEA 67.8 GBP
SIAL269433-100G
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol, Sigma-Aldrich®
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol
Formula: C₅H₄F₈O
MW: 232.07 g/mol
Boiling Pt: 141 °C (1013 hPa)
Melting Pt: < –50 °C
Density: 1.663 g/cm³ (20 °C)
MDL Number: MFCD00039631
CAS Number: 355-80-6
EINECS: 206-593-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR