2'-Bromoacetophenone
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
o-Bromoacetophenone
OS-7606-5G
OS-7606-25G
OS-7606-100G
COBBOS-7606-5GEA
0
GBP
COBBOS-7606-5G
COBBOS-7606-25G
COBBOS-7606-100G
2'-Bromoacetophenone
2'-Bromoacetophenone
Formula:
C₈H₇BrO MW: 199.05 g/mol Boiling Pt: 249 °C (1013 hPa) Melting Pt: 0 °C Density: 1,476 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000067 CAS Number: 2142-69-0 EINECS: 218-398-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...