2-Bromo-5-fluorobenzonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OS-7576-5G
OS-7576-25G
COBBOS-7576-5GEA
0
GBP
COBBOS-7576-5G
COBBOS-7576-25G
2-Bromo-5-fluorobenzonitrile
2-Bromo-5-fluorobenzonitrile
Formula:
C₇H₃BrFN MW: 200.01 g/mol Boiling Pt: 255 °C (1013 hPa) Melting Pt: 89…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00142875 CAS Number: 57381-39-2 EINECS: 260-711-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...