1-(4-Chlorophenyl)-3-methyl-1H-pyrazole

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(4-Chlorophenyl)-3-methyl-1H-pyrazole

HI-1566-5G HI-1566-1G
COBBHI-1566-5GEA 0 GBP
COBBHI-1566-5G COBBHI-1566-1G
1-(4-Chlorophenyl)-3-methyl-1H-pyrazole
1-(4-Chlorophenyl)-3-methyl-1H-pyrazole
Formula: C₁₀H₉ClN₂
MDL Number: MFCD11848681
CAS Number: 27301-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR