Phenol red sodium salt, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxidePhenol red water soluble3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt

M-Clarity™ quality level = MQ200

1.11748.0005EA 71.9 GBP
1.11748.0005
Phenol red sodium salt, Supelco®
Phenol red sodium salt
Formula: C₁₉H₁₃NaO₅S
MW: 376.35 g/mol
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00066901
CAS Number: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397

Order Now

Specification Test Results

Solubility 100 g/l (20 °C)
Melting Point 285 °C
Molar Mass 376.35 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR