Dithizone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,5-DiphenylthiocarbazoneDithizone,for analysis

M-Clarity™ quality level = MQ100

SIALD5130-10GEA 95.2 GBP
SIALD5130-10G SIALD5130-50G
Dithizone, Sigma-Aldrich®
Dithizone
Formula: C₆H₅NHNHCSN=NC₆H₅
MW: 256.33 g/mol
Boiling Pt: 406 °C (1013 hPa)
Melting Pt: 168 °C
MDL Number: MFCD00003025
CAS Number: 60-10-6
EINECS: 200-454-1
Merck Index: 13,03413

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR