(2-Chlorophenyl)acetic acid 98%

Supplier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Chlorophenylacetic acid

OR3987-100G OR3987-25G OR3987-500G
APOSOR3987-100GEA 69.4 GBP
APOSOR3987-100G APOSOR3987-25G APOSOR3987-500G
(2-Chlorophenyl)acetic acid 98%
(2-Chlorophenyl)acetic acid
Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Boiling Pt: 145 °C/5mmHg
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00004317
CAS Number: 2444-36-2
EINECS: 219-482-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR