Barium carbonate 99.99+%
Supplier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
APOSIN1177-25GEA
99.9
GBP
APOSIN1177-25G
Barium carbonate 99.99+%
Barium carbonate
Formula:
BaCO₃ MW: 197.34 g/mol Melting Pt: 1740 °C Density: 4.43 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003448 CAS Number: 513-77-9 EINECS: 208-167-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...