Barium carbonate

Supplier: Roth Carl
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

ROTHP008.1EA 96.98 GBP
ROTHP008.1
Barium carbonate
Barium carbonate
Formula: BaCO₃
MW: 197.34 g/mol
Melting Pt: 1350 °C
Density: 3.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Merck Index: 13,00972

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR