Barium carbonate ≥99.8%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

14341.18EA 32.2 GBP
14341.18 14341.30 14341.A1 14341.A7
Barium carbonate ≥99.8%
Barium carbonate
Formula: BaCO₃
MW: 197.34 g/mol
Boiling Pt: Dec.
Melting Pt: 811 °C
Density: 4.43
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Merck Index: 14,969

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR