α-Bromo-4-iodotoulene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
4-Iodobenzyl bromide, 1-(Bromomethyl)-4-iodobenzene, α-Bromo-p-iodotoulene
018740-5G
018740-25G
FLUO018740-5GEA
0
GBP
FLUO018740-5G
FLUO018740-25G
α-Bromo-4-iodotoulene
α-Bromo-4-iodotoulene
Formula:
C₇H₆BrI MW: 296.93 g/mol Boiling Pt: 115…120 °C (0.6 mmHg) Melting Pt: 80 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00209656 CAS Number: 16004-15-2 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...