1-Bromo-3-methylbutane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 3-Methylbutylbromide

M-Clarity™ quality level = MQ200

124095-100G 124095-500G 124095-5G
SIAL124095-100GEA 55.6 GBP
SIAL124095-100G SIAL124095-500G SIAL124095-5G
1-Bromo-3-methylbutane, Sigma-Aldrich®
1-Bromo-3-methylbutane
Formula: C₅H₁₁Br
MW: 151.05 g/mol
Boiling Pt: 119 °C (1013 hPa)
Melting Pt: –112 °C
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000253
CAS Number: 107-82-4
EINECS: 203-522-9
UN: 2341
ADR: 3,III
Merck Index: 13,05130

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR