Sodium hydroxide 2.05 mol/l (2.05 N) in aqueous solution

Supplier: PROLABO CMP
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Caustic soda

PRLS85692.410EA 0 GBP
PRLS85692.410
Sodium hydroxide 2.05 mol/l (2.05 N) in aqueous solution
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR