Hydrochloric acid 2 mol/l (2 N)

Supplier: PROLABO CMP
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Muriatic acidHydrochloric acid solution

PRLSMC240305.2000EA 0 GBP
PRLSMC240305.2000 PRLSMC240305.5000 PRLSMC240305.1000 PRLSMC240305.5010 PRLSMC240305.9010 PRLSMC240305.9020
Hydrochloric acid 2 mol/l (2 N)
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR