(±)-1-Ethoxy-2-propanol

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

EHERC13309000EA 96.8 GBP
EHERC13309000
(±)-1-Ethoxy-2-propanol
(±)-1-Ethoxy-2-propanol
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 132 °C (1013 hPa)
Melting Pt: –59 °C
Density: 0.9028 g/cm³ (20 °C)
MDL Number: MFCD00067050
CAS Number: 1569-02-4
EINECS: 216-374-5
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR