Anti-GFAP Mouse Monoclonal Antibody [clone: GA-5 ASTRO/789]

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BNUB0898-500 BNUB0898-100
BNUB0898-500EA 487 GBP
BNUB0898-500 BNUB0898-100
Anti-GFAP Mouse Monoclonal Antibody [clone: GA-5 ASTRO/789]
Antibodies
This MAb recognizes a protein of ~50 kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins. GFAP is specifically found in astroglia. GFAP is a very popular marker for localizing benign astrocyte and neoplastic cells of glial origin in the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS.

  • Purified

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: GA-5 ASTRO/789
Conjugation: Purified
Epitope:
Host: Mouse
Reactivity: Chicken, Cow, Human, Mouse, Pig, Rabbit, Rat
Isotype: IgGs
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR