Dithizone ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,5-Diphenylthiocarbazone
, Dithizone,for analysis
A17947.09EA
31.6
GBP
A17947.09
Dithizone ≥98%
Dithizone
Purity: 98%
CAS No.: 60-10-6
Molecular Formula: C13H12N4S
Formula Weight: 256.33
Odour: Odourless
Melting Point: ca 168° dec.
Beilstein Reference: 748838
Merck Reference: 14,3377
MDL No.: MFCD00003025
Formula:
C₆H₅NHNHCSN=NC₆H₅ MW: 256.33 g/mol Boiling Pt: 406 °C (1013 hPa) Melting Pt: 168 °C |
MDL Number:
MFCD00003025 CAS Number: 60-10-6 EINECS: 200-454-1 Merck Index: 13,03413 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...