p-Xylylenediamine for synthesis, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1,4-Bis(aminomethyl)benzene
, 1,4-Phenylenedimethanamine
M-Clarity™ quality level = MQ200
8.41656.0010EA
77.6
GBP
8.41656.0010
p-Xylylenediamine for synthesis, Sigma-Aldrich®
p-Xylylenediamine
Formula:
C₈H₁₂N₂ MW: 136.20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00009821 CAS Number: 539-48-0 EINECS: 208-719-3 UN: 3259 ADR: 8,II |
Specification Test Results
Melting Point | 60 - 64 °C |
Molar Mass | 136.19 g/mol |
Boiling Point | 230 °C (13.33 hPa) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...